Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCB4
Entrez Gene ID
842278
Gene Name
cofactor assembly of complex C
Gene Aliases
AT1G59840, F23H11.16, F23H11_16, cofactor assembly of complex C
Chromosome Location
Chr.1: 22026462 - 22029379 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333869.1 NP_001321983.1 3 - 4 453 77
NM_104677.5 NP_176193.2 3 - 4 500 77
NM_202322.2 NP_974051.1 3 - 4 500 77
GenBank mRNA AK175262.1 - 3 - 4 400 77
AK175368.1 - 3 - 4 400 77
AK221539.1 - 3 - 4 400 77
BT010443.1 - 3 - 4 393 77

View Full Product Details