Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGLU4
Entrez Gene ID
842304
Gene Name
beta glucosidase 4
Gene Aliases
AT1G60090, T2K10.15, T2K10_15, beta glucosidase 4
Chromosome Location
Chr.1: 22155559 - 22158272 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104701.3 NP_176217.2 10 - 11 1078 96

View Full Product Details