Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS34
Entrez Gene ID
842344
Gene Name
vacuolar protein sorting 34
Gene Aliases
AT1G60490, ATVPS34, F8A5.4, F8A5_4, PHOSPATIDYLINOSITOL 3-KINASE, PHOSPHATIDYLINOSITOL 3-KINASE, PI3K, vacuolar protein sorting 34
Chromosome Location
Chr.1: 22285572 - 22290389 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104735.4 NP_176251.1 4 - 5 669 70
GenBank mRNA BT000878.1 - 4 - 5 470 70

View Full Product Details