Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIK3
Entrez Gene ID
842374
Gene Name
NSP-interacting kinase 3
Gene Aliases
AT1G60800, AtNIK3, F8A5.31, F8A5_31, NSP-interacting kinase 3
Chromosome Location
Chr.1: 22383393 - 22387766 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333916.1 NP_001322341.1 2 - 3 461 71
NM_104763.3 NP_176279.1 3 - 4 1145 71
GenBank mRNA AY056294.1 - 3 - 4 836 71
AY150495.1 - 3 - 4 310 71
FJ708666.1 - 3 - 4 310 71

View Full Product Details