Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FED A
Entrez Gene ID
842386
Gene Name
2Fe-2S ferredoxin-like superfamily protein
Gene Aliases
AT1G60950, ATFD2, FD2, FERREDOXIN 2, FERRODOXIN A
Chromosome Location
Chr.1: 22444307 - 22445291 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104775.3 NP_176291.1 1 - 1 767 118
GenBank mRNA AF324706.2 - 1 - 1 554 118
AF326885.1 - 1 - 1 554 118
AK226379.1 - 1 - 1 523 118
AY093034.1 - 1 - 1 560 118
AY128936.1 - 1 - 1 509 118
M35868.1 - 1 - 1 1029 118

View Full Product Details