Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLO6
Entrez Gene ID
842450
Gene Name
Seven transmembrane MLO family protein
Gene Aliases
AT1G61560, ATMLO6, MILDEW RESISTANCE LOCUS O 6, T25B24.9, T25B24_9
Chromosome Location
Chr.1: 22708468 - 22712289 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198352.1 NP_001185281.1 8 - 9 937 64
NM_001198354.1 NP_001185283.1 8 - 9 939 64
NM_001333983.1 NP_001322619.1 8 - 9 1157 64
NM_001333984.1 NP_001319285.1 8 - 9 1102 64
NM_001333985.1 NP_001322617.1 8 - 9 1181 64
NM_001333986.1 NP_001322615.1 8 - 9 1181 64
NM_001333987.1 NP_001322616.1 8 - 9 1181 64
NM_001333988.1 NP_001322614.1 8 - 9 1181 64
NM_001333989.1 NP_001322618.1 8 - 9 1157 64
NM_104836.2 NP_176350.1 8 - 9 1181 64
GenBank mRNA AF369567.2 - 8 - 9 936 64

View Full Product Details