Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAN
Entrez Gene ID
842469
Gene Name
NAD(P)-binding Rossmann-fold superfamily protein
Gene Aliases
AT1G61720, BANYULS, T13M11.8, T13M11_8
Chromosome Location
Chr.1: 22790966 - 22792803 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104854.4 NP_176365.1 1 - 2 178 113
GenBank mRNA DQ446384.1 - 1 - 2 132 113

View Full Product Details