Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY6
Entrez Gene ID
842527
Gene Name
WRKY family transcription factor
Gene Aliases
AT1G62300, ATWRKY6, F19K23.22, F19K23_22
Chromosome Location
Chr.1: 23016569 - 23019494 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104910.3 NP_564792.1 2 - 3 812 140
GenBank mRNA AB493514.1 - 2 - 3 473 140
AF331712.1 - 2 - 3 529 140

View Full Product Details