Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- STM
- Entrez Gene ID
-
842534
- Gene Name
- KNOX/ELK homeobox transcription factor
- Gene Aliases
-
AT1G62360, BUM, BUM1, BUMBERSHOOT, BUMBERSHOOT 1, F24O1.38, F24O1_38, HOMEOBOX PROTEIN SHOOTMERISTEMLESS, SHL, SHOOT MERISTEMLESS, SHOOTLESS, WALDMEISTER, WALDMEISTER 1, WAM, WAM1
- Chromosome Location
-
Chr.1: 23058582 - 23062063 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104916.4 |
NP_176426.1 |
3 - 4 |
1283 |
69 |
View Full Product Details