The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001334038.1 | NP_001321026.1 | 3 - 4 | 958 | 68 |
NM_104934.3 | NP_176444.1 | 3 - 4 | 958 | 68 | |
GenBank mRNA | AF370136.1 | - | 3 - 4 | 869 | 68 |
AY113858.1 | - | 3 - 4 | 780 | 68 |