Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BETA-VPE
Entrez Gene ID
842569
Gene Name
beta vacuolar processing enzyme
Gene Aliases
AT1G62710, BETAVPE, F23N19.7, F23N19_7, beta vacuolar processing enzyme
Chromosome Location
Chr.1: 23223874 - 23226983 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104948.4 NP_176458.1 3 - 4 652 90
GenBank mRNA AF367254.1 - 3 - 4 556 90
AY059156.1 - 3 - 4 526 90

View Full Product Details