Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXPA18
Entrez Gene ID
842601
Gene Name
expansin A18
Gene Aliases
AT1G62980, ATEXP18, ATEXPA18, ATHEXP ALPHA 1.25, EXP18, EXPANSIN 18, F16P17.14, F16P17_14, expansin A18
Chromosome Location
Chr.1: 23331563 - 23333246 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104976.5 NP_176486.1 1 - 2 307 90
GenBank mRNA AF332444.1 - 1 - 2 142 90
AK227678.1 - 1 - 2 194 90
BT006412.1 - 1 - 2 142 90

View Full Product Details