Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXPA18
Entrez Gene ID
842601
Gene Name
expansin A18
Gene Aliases
AT1G62980, ATEXP18, ATEXPA18, ATHEXP ALPHA 1.25, EXP18, EXPANSIN 18, F16P17.14, F16P17_14, expansin A18
Chromosome Location
Chr.1: 23331563 - 23333246 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104976.5 NP_176486.1 2 - 3 624 114
GenBank mRNA AF332444.1 - 2 - 3 459 114
AK227678.1 - 2 - 3 511 114
BT006412.1 - 2 - 3 459 114

View Full Product Details