The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_104990.4 | NP_176500.1 | 1 - 2 | 690 | 143 |
GenBank mRNA | AB195671.1 | - | 1 - 2 | 276 | 143 |
BT004076.1 | - | 1 - 2 | 366 | 143 | |
BT005121.1 | - | 1 - 2 | 276 | 143 |