Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMO1-1
Entrez Gene ID
826819
Gene Name
sterol-4alpha-methyl oxidase 1-1
Gene Aliases
AT4G12110, ATSMO1, ATSMO1-1, F16J13.180, F16J13_180, STEROL 4-ALPHA-METHYL-OXIDASE 1, sterol-4alpha-methyl oxidase 1-1
Chromosome Location
Chr.4: 7254029 - 7256406 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117281.3 NP_192948.1 2 - 3 833 81
GenBank mRNA AY125499.1 - 2 - 3 811 81
BT000546.1 - 2 - 3 665 81

View Full Product Details