Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORP2B
Entrez Gene ID
826858
Gene Name
OSBP(oxysterol binding protein)-related protein 2B
Gene Aliases
AT4G12460, OSBP(oxysterol binding protein)-related protein 2B, T1P17.50, T1P17_50
Chromosome Location
Chr.4: 7391208 - 7396511 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084907.1 NP_001078376.1 6 - 7 1156 92
NM_001340775.1 NP_001327964.1 6 - 7 2450 92
NM_001340776.1 NP_001327965.1 7 - 8 2160 92
NM_001340777.1 NP_001327966.1 6 - 7 1636 92
NM_001340778.1 NP_001327967.1 6 - 7 1613 92
NM_001340779.1 NP_001327968.1 6 - 7 1613 92
NM_001340780.1 NP_001327962.1 6 - 7 1567 92
NM_001340781.1 NP_001327963.1 5 - 6 1367 92
NM_117316.3 NP_192983.2 6 - 7 1636 92
GenBank mRNA AY140023.1 - 6 - 7 1154 92
BT008852.1 - 3 - 4 466 92

View Full Product Details