Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORP2B
Entrez Gene ID
826858
Gene Name
OSBP(oxysterol binding protein)-related protein 2B
Gene Aliases
AT4G12460, OSBP(oxysterol binding protein)-related protein 2B, T1P17.50, T1P17_50
Chromosome Location
Chr.4: 7391208 - 7396511 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084907.1 NP_001078376.1 9 - 10 1616 90
NM_001340775.1 NP_001327964.1 9 - 10 2916 90
NM_001340776.1 NP_001327965.1 10 - 11 2626 90
NM_001340777.1 NP_001327966.1 8 - 9 2225 90
NM_001340778.1 NP_001327967.1 8 - 9 2202 90
NM_001340779.1 NP_001327968.1 9 - 10 2079 90
NM_001340780.1 NP_001327962.1 9 - 10 2033 90
NM_001340781.1 NP_001327963.1 8 - 9 1833 90
NM_117316.3 NP_192983.2 9 - 10 2102 90
GenBank mRNA AY140023.1 - 9 - 10 1614 90
BT008852.1 - 6 - 7 926 90

View Full Product Details