Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PTAC12
- Entrez Gene ID
-
818029
- Gene Name
- plastid transcriptionally active 12
- Gene Aliases
-
AT2G34640, HEMERA, HMR, T31E10.2, T31E10_2, TAC12, plastid transcriptionally active 12
- Chromosome Location
-
Chr.2: 14581835 - 14584797 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129018.4 |
NP_181011.2 |
3 - 4 |
1040 |
70 |
View Full Product Details