Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCC2
Entrez Gene ID
818031
Gene Name
multidrug resistance-associated protein 2
Gene Aliases
ABC TRANSPORTER, AT2G34660, ATMRP2, ATP-binding cassette C2, Arabidopsis thaliana ATP-binding cassette C2, AtABCC2, EST4, MRP2, T29F13.13, T29F13_13, multidrug resistance-associated protein 2
Chromosome Location
Chr.2: 14602470 - 14612565 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202746.1 NP_001189675.1 22 - 23 3210 125
NM_001336522.1 NP_001324371.1 16 - 17 2183 125
NM_129020.3 NP_181013.1 21 - 22 3475 125

View Full Product Details