Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MEE27
- Entrez Gene ID
-
818053
- Gene Name
- Transcription factor jumonji (jmj) family protein / zinc finger (C5HC2 type) family protein
- Gene Aliases
-
AT2G34880, F19I3.11, F19I3_11, JMJ15, Jumonji domain-containing protein 15, PKDM7C, maternal effect embryo arrest 27
- Chromosome Location
-
Chr.2: 14711880 - 14716634 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129041.1 |
NP_181034.1 |
5 - 6 |
691 |
93 |
View Full Product Details