Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMB1
Entrez Gene ID
818055
Gene Name
Transcription factor GTE6
Gene Aliases
AT2G34900, F19I3.13, F19I3_13, GLOBAL TRANSCRIPTION FACTOR GROUP E1, GTE01, GTE1, IMBIBITION-INDUCIBLE 1
Chromosome Location
Chr.2: 14723013 - 14725684 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129043.5 NP_181036.2 2 - 3 269 72
NM_201873.2 NP_973602.1 2 - 3 216 72
GenBank mRNA AK230434.1 - 2 - 3 231 72
AY180100.1 - 2 - 3 147 72
BT026469.1 - 2 - 3 147 72

View Full Product Details