Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMB1
Entrez Gene ID
818055
Gene Name
Transcription factor GTE6
Gene Aliases
AT2G34900, F19I3.13, F19I3_13, GLOBAL TRANSCRIPTION FACTOR GROUP E1, GTE01, GTE1, IMBIBITION-INDUCIBLE 1
Chromosome Location
Chr.2: 14723013 - 14725684 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129043.5 NP_181036.2 4 - 5 566 110
NM_201873.2 NP_973602.1 4 - 5 513 110
GenBank mRNA AK230434.1 - 4 - 5 528 110
AY180100.1 - 4 - 5 444 110
BT026469.1 - 4 - 5 444 110

View Full Product Details