Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SETH1
Entrez Gene ID
818062
Gene Name
phosphatidylinositolglycan synthase family protein
Gene Aliases
AT2G34980, F19I3.21, F19I3_21
Chromosome Location
Chr.2: 14748877 - 14749878 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129050.2 NP_181043.1 1 - 1 962 85

View Full Product Details