Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TET10
Entrez Gene ID
842632
Gene Name
tetraspanin10
Gene Aliases
AT1G63260, F9N12.12, F9N12_12, tetraspanin10
Chromosome Location
Chr.1: 23466700 - 23469289 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036145.3 NP_001031222.1 5 - 6 650 125
NM_001334075.1 NP_001320678.1 5 - 6 627 125
NM_001334076.1 NP_001320677.1 5 - 6 669 125
NM_001334077.1 NP_001320676.1 6 - 7 568 125
NM_105005.5 NP_176515.3 5 - 6 650 125
NM_202348.3 NP_974077.1 5 - 6 650 125
GenBank mRNA BT011569.1 - 5 - 6 511 125
BT012253.1 - 5 - 6 325 125

View Full Product Details