The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_105025.4 | NP_564813.1 | 4 - 5 | 469 | 111 |
GenBank mRNA | AK227611.1 | - | 4 - 5 | 399 | 111 |
AY086991.1 | - | 4 - 5 | 313 | 111 | |
BT003059.1 | - | 4 - 5 | 297 | 111 |