Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YDA
Entrez Gene ID
842674
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT1G63700, EMB71, EMBRYO DEFECTIVE 71, F24D7.11, F24D7_11, MAP KINASE KINASE KINASE 4, MAPKKK4, YODA
Chromosome Location
Chr.1: 23624627 - 23630073 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334122.1 NP_001319310.1 6 - 7 2109 84
NM_105047.2 NP_176557.1 5 - 6 1659 84
GenBank mRNA AY357947.1 - 5 - 6 1659 84

View Full Product Details