Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RBOH F
- Entrez Gene ID
-
842710
- Gene Name
- respiratory burst oxidase protein F
- Gene Aliases
-
ARABIDOPSIS THALIANA RESPIRATORY BURST OXIDASE HOMOLOG F, ARABIDOPSIS THALIANA RESPIRATORY BURST OXIDASE HOMOLOG F, AT1G64060, ATRBOH F, ATRBOHF, CYTOCHROME B245 BETA CHAIN HOMOLOG RBOHAP108, F22C12.18, F22C12_18, RBOHAP108, RBOHF, RESPIRATORY BURST OXIDASE PROTEIN F, respiratory burst oxidase protein F
- Chromosome Location
-
Chr.1: 23769774 - 23776984 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105079.3 |
NP_564821.1 |
7 - 8 |
2380 |
93 |
GenBank mRNA |
AF015301.1 |
- |
7 - 8 |
2379 |
93 |
View Full Product Details