Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCF3
Entrez Gene ID
842763
Gene Name
general control non-repressible 3
Gene Aliases
AT1G64550, ATGCN3, ATP-binding cassette F3, AtABCF3, AtGCN20, F1N19.28, F1N19_28, GCN20, GCN3, SCORD5, general control non-repressible 20, general control non-repressible 3, susceptible to coronatine-deficient Pst DC3000 5
Chromosome Location
Chr.1: 23968750 - 23973614 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105130.4 NP_176636.1 3 - 4 709 151
GenBank mRNA BT002015.1 - 3 - 4 651 151
BT010544.1 - 3 - 4 609 151

View Full Product Details