Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WNK10
Entrez Gene ID
842771
Gene Name
with no lysine (K) kinase 10
Gene Aliases
AT1G64630, ATWNK10, F1N19.20, F1N19_20, WITH NO LYSINE KINASE 10, with no lysine (K) kinase 10
Chromosome Location
Chr.1: 24019490 - 24022310 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105138.5 NP_176644.2 6 - 7 1525 106
GenBank mRNA AB493519.1 - 6 - 7 1095 106
AY081307.1 - 6 - 7 1516 106
BT010383.1 - 6 - 7 1095 106

View Full Product Details