Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PnsB2
Entrez Gene ID
842785
Gene Name
NDH-dependent cyclic electron flow 1
Gene Aliases
AT1G64770, F13O11.8, F13O11_8, NAD(P)H DEHYDROGENASE SUBUNIT 45, NDF2, NDH-dependent cyclic electron flow 1, NDH45, Photosynthetic NDH subcomplex B 2
Chromosome Location
Chr.1: 24057063 - 24059621 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036156.2 NP_001031233.1 1 - 2 519 62
NM_001198383.1 NP_001185312.1 1 - 2 895 62
NM_105151.4 NP_564840.1 1 - 2 661 62
GenBank mRNA AY045871.1 - 1 - 2 480 62
AY085928.1 - 1 - 2 519 62
AY113953.1 - 1 - 2 409 62

View Full Product Details