Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
G-TMT
Entrez Gene ID
842805
Gene Name
gamma-tocopherol methyltransferase
Gene Aliases
AT1G64970, F13O11.27, F13O11_27, GAMMA-TOCOPHEROL METHYLTRANSFERASE, TMT1, VITAMIN E DEFICIENT 4, VTE4, gamma-tocopherol methyltransferase
Chromosome Location
Chr.1: 24134085 - 24136109 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105171.3 NP_176677.1 3 - 4 953 70
GenBank mRNA AY049258.1 - 3 - 4 904 70
AY090280.1 - 3 - 4 837 70

View Full Product Details