Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAC4
Entrez Gene ID
842833
Gene Name
plastid transcriptionally active 4
Gene Aliases
AT1G65260, T8F5.2, T8F5_2, VESICLE-INDUCING PROTEIN IN PLASTIDS 1, VIPP1, plastid transcriptionally active 4
Chromosome Location
Chr.1: 24236088 - 24240728 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105199.3 NP_564846.1 1 - 2 384 100
GenBank mRNA AY045845.1 - 1 - 2 152 100
AY117153.1 - 1 - 2 143 100

View Full Product Details