Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTO1
Entrez Gene ID
821292
Gene Name
Pyridoxal phosphate (PLP)-dependent transferases superfamily protein
Gene Aliases
A. THALIANA CYSTATHIONINE GAMMA-SYNTHASE 1, A. thaliana cystathionine gamma-synthetase 1, AT3G01120, ATCYS1, AtCGS1, AtCYS1, CGS, CGS1, CYSTATHIONINE GAMMA-SYNTHASE, CYSTATHIONINE GAMMA-SYNTHASE 1, CYSTATHIONINE-GAMMA-SYNTHASE, METHIONINE OVERACCUMULATION 1, T4P13.19, T4P13_19
Chromosome Location
Chr.3: 38759 - 42159 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_110977.3 NP_186761.1 6 - 7 1457 81
GenBank mRNA AY094438.1 - 6 - 7 1195 81
BT002753.1 - 6 - 7 1163 81
U83500.1 - 6 - 7 1183 81

View Full Product Details