Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTB1
Entrez Gene ID
821129
Gene Name
polypyrimidine tract-binding protein 1
Gene Aliases
AT3G01150, ATPTB1, POLYPYRIMIDINE TRACT-BINDING, POLYPYRIMIDINE TRACT-BINDING PROTEIN 1, PTB, T4P13.16, T4P13_16, polypyrimidine tract-binding protein 1
Chromosome Location
Chr.3: 51496 - 54544 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337324.1 NP_001327637.1 2 - 3 630 85
NM_110980.4 NP_186764.1 2 - 3 630 85
GenBank mRNA AK316897.1 - 2 - 3 478 85

View Full Product Details