Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VDAC1
Entrez Gene ID
820914
Gene Name
voltage dependent anion channel 1
Gene Aliases
5076, ARABIDOPSIS THALIANA VOLTAGE DEPENDENT ANION CHANNEL 1, AT3G01280, ATVDAC1, T22N4.9, T22N4_9, voltage dependent anion channel 1
Chromosome Location
Chr.3: 85551 - 88070 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_110994.4 NP_186777.1 5 - 6 734 112
GenBank mRNA AY037217.1 - 5 - 6 648 112
AY113169.1 - 5 - 6 531 112

View Full Product Details