Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAP74
Entrez Gene ID
826874
Gene Name
transcription initiation factor IIF subunit alpha RAP74
Gene Aliases
AT4G12610, ATRAP74, T1P17.200, T1P17_200
Chromosome Location
Chr.4: 7454722 - 7458422 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340791.1 NP_001319913.1 4 - 5 594 132
NM_001340792.1 NP_001319914.1 4 - 5 577 132

View Full Product Details