The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001340813.1 | NP_001329616.1 | 4 - 5 | 883 | 84 |
NM_117360.8 | NP_193027.5 | 4 - 5 | 926 | 84 |