Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MEE57
- Entrez Gene ID
-
826994
- Gene Name
- DNA (cytosine-5-)-methyltransferase family protein
- Gene Aliases
-
AT4G13610, T6G15.160, T6G15_160, maternal effect embryo arrest 57
- Chromosome Location
-
Chr.4: 7915022 - 7921231 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117435.1 |
NP_193097.1 |
5 - 6 |
2785 |
142 |
View Full Product Details