Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR2
Entrez Gene ID
827000
Gene Name
pinoresinol reductase 2
Gene Aliases
AT4G13660, ATPRR2, F18A5.50, F18A5_50, PSEUDO-RESPONSE REGULATOR 2, pinoresinol reductase 2
Chromosome Location
Chr.4: 7945683 - 7948443 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340871.1 NP_001319922.1 3 - 4 1062 134
NM_117440.3 NP_193102.1 2 - 3 962 134
GenBank mRNA BT002882.1 - 2 - 3 713 134
BT004406.1 - 2 - 3 585 134

View Full Product Details