Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NOV
- Entrez Gene ID
-
827009
- Gene Name
- Histidine kinase-, DNA gyrase B-, and HSP90-like ATPase family protein
- Gene Aliases
-
AT4G13750, EMB2597, EMBRYO DEFECTIVE 2597, F18A5.140, F18A5_140, NO VEIN
- Chromosome Location
-
Chr.4: 7975064 - 7987687 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117449.4 |
NP_193111.2 |
7 - 8 |
3975 |
147 |
View Full Product Details