Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AHL20
Entrez Gene ID
827093
Gene Name
AT-hook motif nuclear-localized protein 20
Gene Aliases
AT-hook motif nuclear-localized protein 20, AT4G14465
Chromosome Location
Chr.4: 8320538 - 8322352 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117526.4 NP_567432.1 1 - 1 1434 103
GenBank mRNA AK118702.1 - 1 - 1 1342 103
AK227935.1 - 1 - 1 1342 103

View Full Product Details