Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOI1
- Entrez Gene ID
-
827142
- Gene Name
- Pentatricopeptide repeat (PPR) superfamily protein
- Gene Aliases
-
AT4G14850, DL3465W, FCAALL.335, MEF11, lovastatin insensitive 1, mitochondrial RNA editing factor 11
- Chromosome Location
-
Chr.4: 8513807 - 8516360 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117571.5 |
NP_193221.3 |
1 - 2 |
1728 |
69 |
GenBank mRNA |
AK227954.1 |
- |
1 - 2 |
1592 |
69 |
View Full Product Details