Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG2
Entrez Gene ID
827183
Gene Name
SET domain protein 2
Gene Aliases
AT4G15180, ATXR3, DL3635W, FCAALL.214, SET domain protein 2
Chromosome Location
Chr.4: 8651406 - 8662587 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117606.5 NP_193253.4 12 - 13 6063 65

View Full Product Details