Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RH1
Entrez Gene ID
827266
Gene Name
RNA helicase 1
Gene Aliases
AT4G15850, ATRH1, DL3965W, FCAALL.401, RNA helicase 1
Chromosome Location
Chr.4: 9001174 - 9004734 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117677.7 NP_193320.6 3 - 4 866 69

View Full Product Details