Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TS1
Entrez Gene ID
827268
Gene Name
terpene synthase 1
Gene Aliases
AT4G15870, ATTS1, DL3975C, FCAALL.405, terpene synthase 1
Chromosome Location
Chr.4: 9008372 - 9010947 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117679.2 NP_193322.2 4 - 5 1148 92
GenBank mRNA Y11188.1 - - 1148 92

View Full Product Details