Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATOEP16-2
Entrez Gene ID
827308
Gene Name
Mitochondrial import inner membrane translocase subunit Tim17/Tim22/Tim23 family protein
Gene Aliases
AT4G16160, ATOEP16-S, DL4120W, FCAALL.207
Chromosome Location
Chr.4: 9157471 - 9158933 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117712.2 NP_567488.1 3 - 4 267 77
NM_179063.2 NP_849394.1 3 - 4 329 77
GenBank mRNA AK229661.1 - 3 - 4 277 77
AY088334.1 - 3 - 4 268 77

View Full Product Details