Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHYD
Entrez Gene ID
827319
Gene Name
phytochrome D
Gene Aliases
AT4G16250, DL4165C, FCAALL.323, PHYTOCHROME D, phytochrome D
Chromosome Location
Chr.4: 9195247 - 9199840 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117721.2 NP_193360.1 2 - 3 3340 85

View Full Product Details