Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPK1
Entrez Gene ID
827328
Gene Name
guanyl-nucleotide exchange factors;GTPase binding;GTP binding protein
Gene Aliases
AT4G16340, DL4200C, FCAALL.346, SPIKE1
Chromosome Location
Chr.4: 9228185 - 9241629 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341097.1 NP_001319958.1 6 - 7 869 145
NM_117729.8 NP_193367.7 5 - 6 821 145
GenBank mRNA AF465831.1 - 5 - 6 754 145

View Full Product Details