Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OPT3
Entrez Gene ID
827332
Gene Name
oligopeptide transporter
Gene Aliases
AT4G16370, ATOPT3, DL4215C, FCAALL.72, oligopeptide transporter
Chromosome Location
Chr.4: 9247302 - 9250343 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117732.7 NP_567493.5 4 - 5 1689 95

View Full Product Details