Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACX1
Entrez Gene ID
827381
Gene Name
acyl-CoA oxidase 1
Gene Aliases
AT4G16760, ATACX1, DL4405C, FCAALL.119, acyl-CoA oxidase 1
Chromosome Location
Chr.4: 9424534 - 9428895 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203815.1 NP_001190744.1 13 - 14 1950 82
NM_117778.8 NP_567513.4 13 - 14 1988 82
GenBank mRNA AF057044.1 - 13 - 14 1859 82
AY058849.2 - 13 - 14 1916 82
BT001067.1 - 13 - 14 1783 82

View Full Product Details